MilliporeSigma
  • FT-IR, FT-Raman and UV-Vis spectra and DFT calculations of 3-cyano-4-methylcoumarin.
FT-IR, FT-Raman and UV-Vis spectra and DFT calculations of 3-cyano-4-methylcoumarin.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2012-08-16)
N Udaya Sri, K Chaitanya, M V S Prasad, V Veeraiah, A Veeraiah
ABSTRACT
The vibrational and electronic spectra of 3-cyano-4-methylcoumarin (3C4MC) are reported and discussed. In this work the structural properties, vibrational frequencies and electronic spectra of 3C4MC have been investigated extensively using density functional theory (DFT) employing B3LYP exchange correlation with the normal basis level 6-31G(d,p). NBO and HOMO, LUMO analysis has been carried out. The geometries and normal modes of vibrations obtained from B3LYP calculations are in good agreement with the experimentally observed data.