MilliporeSigma
  • Talin-1 correlates with reduced invasion and migration in human hepatocellular carcinoma cells.
Talin-1 correlates with reduced invasion and migration in human hepatocellular carcinoma cells.
Asian Pacific journal of cancer prevention : APJCP (2014-04-26)
Kun-Peng Fang, Jian-Lin Zhang, Yan-Hong Ren, Ye-Ben Qian
ABSTRACT
Talin-1 is a cytoskeleton protein that participates in cell migration and plays a role in tumor formation, migration, and metastasis in different types of cancer. Chinese investigators have observed that the levels of Talin-1 protein and mRNA expression in HCC tissues are significantly lower than in the adjacent non-cancerous tissue. However, Japanese investigators have reported that Talin-1 is upregulated in HCC. Tln2 as homologous gene of Tln-1, which encodes a very similar protein, but the role of Talin-2 is very little known in primary liver cancer (PLC). We investigated whether the expression of Talin-1 in PLC may be associated with the histological subtype as well as the role of Talin-1 in tumor cell invasion and migration using human hepatocellular carcinoma cell lines. We measured the mRNA expression levels of Talin-1 and Talin-2 in five human liver cancer cell lines and normal human liver cell (LO2 cell line) by real-time PCR and the protein expression levels of Talin-1 by Western blot. Migration and invasion of the cells were assessed using transwell assays and cell scratch experiments, respectively, and proliferation was assessed by soft AGAR colony formation. Talin-1 and Talin-2 expression differed significantly between the five human liver cancer cell lines and LO2 cell line (p<0.05). Compared with the LO2 cell line, the invasion and migration capabilities of the five cancer cell lines differed significantly (p<0.05). Similarly, the colony-forming ability differed (p<0.05). High levels of Talin-1 expression are correlated with reduced invasion and migration as well as decreased malignancy in human liver cancer cell lines; the suppression of Talin-1 promotes invasion and migration. In addition, Talin-2 may be correlated with invasion and migration in human hepatocellular carcinoma.