MilliporeSigma
  • Non-IG aberrations of FOXP1 in B-cell malignancies lead to an aberrant expression of N-truncated isoforms of FOXP1.
Non-IG aberrations of FOXP1 in B-cell malignancies lead to an aberrant expression of N-truncated isoforms of FOXP1.
PloS one (2014-01-15)
Leila Rouhigharabaei, Julio Finalet Ferreiro, Thomas Tousseyn, Jo-Anne van der Krogt, Natalie Put, Eugenia Haralambieva, Carlos Graux, Brigitte Maes, Carmen Vicente, Peter Vandenberghe, Jan Cools, Iwona Wlodarska
ABSTRACT
The transcription factor FOXP1 is implicated in the pathogenesis of B-cell lymphomas through chromosomal translocations involving either immunoglobulin heavy chain (IGH) locus or non-IG sequences. The former translocation, t(3;14)(p13;q32), results in dysregulated expression of FOXP1 juxtaposed with strong regulatory elements of IGH. Thus far, molecular consequences of rare non-IG aberrations of FOXP1 remain undetermined. Here, using molecular cytogenetics and molecular biology studies, we comprehensively analyzed four lymphoma cases with non-IG rearrangements of FOXP1 and compared these with cases harboring t(3;14)(p13;q32)/IGH-FOXP1 and FOXP1-expressing lymphomas with no apparent structural aberrations of the gene. Our study revealed that non-IG rearrangements of FOXP1 are usually acquired during clinical course of various lymphoma subtypes, including diffuse large B cell lymphoma, marginal zone lymphoma and chronic lymphocytic leukemia, and correlate with a poor prognosis. Importantly, these aberrations constantly target the coding region of FOXP1, promiscuously fusing with coding and non-coding gene sequences at various reciprocal breakpoints (2q36, 10q24 and 3q11). The non-IG rearrangements of FOXP1, however, do not generate functional chimeric genes but commonly disrupt the full-length FOXP1 transcript leading to an aberrant expression of N-truncated FOXP1 isoforms (FOXP1(NT)), as shown by QRT-PCR and Western blot analysis. In contrast, t(3;14)(p13;q32)/IGH-FOXP1 affects the 5' untranslated region of FOXP1 and results in overexpress the full-length FOXP1 protein (FOXP1(FL)). RNA-sequencing of a few lymphoma cases expressing FOXP1(NT) and FOXP1(FL) detected neither FOXP1-related fusions nor FOXP1 mutations. Further bioinformatic analysis of RNA-sequencing data retrieved a set of genes, which may comprise direct or non-direct targets of FOXP1(NT), potentially implicated in disease progression. In summary, our findings point to a dual mechanism through which FOXP1 is implicated in B-cell lymphomagenesis. We hypothesize that the primary t(3;14)(p13;q32)/IGH-FOXP1 activates expression of the FOXP1(FL) protein with potent oncogenic activity, whereas the secondary non-IG rearrangements of FOXP1 promote expression of the FOXP1(NT) proteins, likely driving progression of disease.
Sigma-Aldrich
Anti-FOXP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.