MilliporeSigma
Formal syntheses of (+/-)-pinnaic acid and (+/-)-halichlorine.
Organic letters (2005-12-03)
Rodrigo B Andrade, Stephen F Martin
ABSTRACT
[chemical reaction: see text]. Concise formal syntheses of marine alkaloids (+/-)-pinnaic acid (1) and (+/-)-halichlorine (2) have been accomplished from a common intermediate. The syntheses illustrate the utility of selective olefin cross metathesis methodologies for the elaboration of advanced synthetic intermediates in complex molecule synthesis.