MilliporeSigma
  • Synthesis of a tritium-labeled, fipronil-based, highly potent, photoaffinity probe for the GABA receptor.
Synthesis of a tritium-labeled, fipronil-based, highly potent, photoaffinity probe for the GABA receptor.
The Journal of organic chemistry (2003-10-11)
Robert E Sammelson, John E Casida
ABSTRACT
3-[4-[1-(2,6-dichloro-4-trifluoromethylphenyl)pyrazolo]]-3-(trifluoromethyl)diazirine is a fipronil-based (i.e. fiprole), high-affinity probe for the GABA receptor. For synthesis of the tritium-labeled version of this trifluoromethyldiazirinylfiprole ([(3)H]TDF) the required intermediate, 3-[4-[1-(2,6-dichloro-3-iodo-4-trifluoromethylphenyl)-5-iodopyrazolo]]-3-(trifluoromethyl)diazirine, was prepared in 10 steps from pyrazole and 3,5-dichloro-4-fluorobenzotrifluoride. One of the key transformations was lithiation and subsequent iodination of the 4-(2,2,2-trifluoro-1-hydroxyethyl)pyrazole intermediate. The last step involved reduction of the diiodofiprole with tritium, Pd/C, and triethylamine in ethyl acetate and afforded [(3)H]TDF with a specific activity of 15 Ci/mmol and 99% radiopurity.