MilliporeSigma
  • Diclofenac sodium multisource prolonged release tablets--a comparative study on the dissolution profiles.
Diclofenac sodium multisource prolonged release tablets--a comparative study on the dissolution profiles.
Journal of pharmaceutical and biomedical analysis (2005-03-31)
Paola Bertocchi, Eleonora Antoniella, Luisa Valvo, Stefano Alimonti, Adriana Memoli
ABSTRACT
The aim of this work was to compare the dissolution behaviour of six diclofenac sodium prolonged release tablets of different brands obtained from the national market. The formulations contain the same amount of drug substance but different types and/or amount of excipients. The influence of these differences in formulation on the release characteristics of the dosage forms was evaluated on the European Pharmacopoeia apparatus 2 (paddle) employing eight different dissolution media in the pH range 1.2-8. Friability and hardness were tested too according to the European Pharmacopoeia. Dissolution profiles obtained from the studied formulations showed that the release characteristics vary considerably among different manufacturers and that even identical formulations show rather dissimilar release profiles in all the studied media. Use of both SIF without pancreatin and SIF without pancreatin containing 1% (w/v) Tween 20 resulted in strong discrimination among products. A correlation between friability and hardness and in vitro dissolution was evidenced for two formulations having identical excipient composition.
Sigma-Aldrich
Magnesium stearate, technical grade
Pricing and availability is not currently available.