MilliporeSigma
  • Reactions between Grignard reagents and heterocyclic N-oxides: stereoselective synthesis of substituted pyridines, piperidines, and piperazines.
Reactions between Grignard reagents and heterocyclic N-oxides: stereoselective synthesis of substituted pyridines, piperidines, and piperazines.
Organic & biomolecular chemistry (2010-11-16)
Hans Andersson, Roger Olsson, Fredrik Almqvist
ABSTRACT
In this perspective we discuss the recent developments of stereoselective synthesis of substituted pyridines, piperidines, and piperazines from cheap and commercially readily available starting materials. Pyridine N-oxides and pyrazine N-oxides are reacted with alkyl, aryl, alkynyl and vinyl Grignard reagents to give a diverse set of heterocycles in high yields. Optically active substituted piperazines are obtained by an asymmetric reaction from pyrazine N-oxides using sparteine as chiral ligand. In addition, a stereoselective synthesis of dienal-oximes from the reaction between pyridine N-oxides and Grignard reagents is presented, which results in a useful intermediate for the synthesis of a diverse set of compounds.