MilliporeSigma
  • MicroRNA-645, up-regulated in human adencarcinoma of gastric esophageal junction, inhibits apoptosis by targeting tumor suppressor IFIT2.
MicroRNA-645, up-regulated in human adencarcinoma of gastric esophageal junction, inhibits apoptosis by targeting tumor suppressor IFIT2.
BMC cancer (2014-09-02)
Xiaoshan Feng, Ying Wang, Zhikun Ma, Ruina Yang, Shuo Liang, Mengxi Zhang, Shiyuan Song, Shuoguo Li, Gang Liu, Daiming Fan, Shegan Gao
ABSTRACT
An increasing body of evidence indicates that miRNAs have a critical role in carcinogenesis and cancer progression; however, the role of miRNAs in the tumorigenesis of adencarcinoma of gastric esophageal junction (AGEJ) remains largely unclear. The SGC7901 and BGC-823 gastric cancer cell lines were used. The expressions of miR-645 and IFIT2 (Interferon-induced protein with tetratricopeptide repeats 2) were examined by qRT-PCR, The expressions of IFIT2 was examined by western blotting and immunohistochemistry assay. The cell apoptosis was determined by FACS. MiR-645 inhibitor, mimics and plasmid-IFIT2 transfections were performed to study the loss- and gain-function. Caspase-3/7 activity was examined by caspase-3/7 assay. In the present study, we have reported an increased expression of miR-645 in AGEJ clinical specimens compared with paired non-cancerous tissues. We also observed a significant miR-645 up-regulation in two gastric cancer (GC) cell lines, SGC7901 and BGC-823, which were used as cell models because there was no available AGEJ cell lines established to date. We found that inhibition of miR-645 could sensitize dramatically SGC7901 and BGC-823 cells to both serum starvation- and chemotherapeutic drug-induced apoptosis by up-regulating IFIT2, a mediator of apoptosis via a mitochondrial pathway, with a potential binding site for miR-645 in its mRNA's 3'UTR. Further investigation exhibited that IFIT2 expression decreases in SGC7901 and BGC-823 cells and AGEJ tissues. IFIT2 ectopic expression leads to promotion of cell apoptosis, indicating that IFIT2 may function as a suppressor in the development of AGEJ. Furthermore, inhibition of miR-645 induces up-regulation of IFIT2 and increased caspase-3/7 activity compared with control groups. Our data suggest that miR-645 functions as an oncogene in human AGEJ by, at least partially through, targeting IFIT2.