MilliporeSigma
  • Preparation of modified peptides: direct conversion of α-amino acids into β-amino aldehydes.
Preparation of modified peptides: direct conversion of α-amino acids into β-amino aldehydes.
Organic & biomolecular chemistry (2012-05-05)
Carlos J Saavedra, Alicia Boto, Rosendo Hernández
ABSTRACT
A direct method for the transformation of α-amino acids into β-amino aldehydes was developed, and applied to the modification of the C-terminal residue of peptides. The method takes place in good yields and under mild conditions. The application of this methodology to the preparation of small peptides with γ-amino alcohol units, which are precursors of analogues of peptaibol antibiotics, is also described.