MilliporeSigma
  • Fast gas chromatography characterisation of purified semiochemicals from essential oils of Matricaria chamomilla L. (Asteraceae) and Nepeta cataria L. (Lamiaceae).
Fast gas chromatography characterisation of purified semiochemicals from essential oils of Matricaria chamomilla L. (Asteraceae) and Nepeta cataria L. (Lamiaceae).
Journal of chromatography. A (2008-10-28)
Stéphanie Heuskin, Bruno Godin, Pascal Leroy, Quentin Capella, Jean-Paul Wathelet, François Verheggen, Eric Haubruge, Georges Lognay
ABSTRACT
The chemical composition of Matricaria chamomilla L. and Nepeta cataria L. essential oils was determined by GC-MS on an apolar stationary phase by comparison of the characteristic fragmentation patterns with those of the Wiley 275L database. The GC-MS chromatograms were compared with those obtained by fast GC equipped with a direct resistively heated column (Ultra Fast Module 5% phenyl, 5 mx 0.1 mm, 0.1 microm film thickness). Analytical conditions were optimised to reach a good peak resolution (split ratio=1:100), with analysis time lower than 5 min versus 35-45 min required by conventional GC-MS. The fast chromatographic method was completely validated for the analysis of mono- and sesquiterpene compounds. Essential oils were then fractionated by column chromatography packed with silica gel. Three main fractions with high degree of purity in E-beta-farnesene were isolated from the oil of M. chamomilla. One fraction enriched in (Z,E)-nepetalactone and one enriched in beta-caryophyllene were obtained from the oil of N. cataria. These semiochemical compounds could act as attractants of aphid's predators and parasitoids.
Sigma-Aldrich
Catalase from bovine liver, aqueous suspension, 10,000-40,000 units/mg protein