MilliporeSigma
  • Internal Lewis acid assisted ureas: tunable hydrogen bond donor catalysts.
Internal Lewis acid assisted ureas: tunable hydrogen bond donor catalysts.
Chemical communications (Cambridge, England) (2012-12-01)
David M Nickerson, Veronica V Angeles, Tyler J Auvil, Sonia S So, Anita E Mattson
ABSTRACT
The strategic incorporation of internal Lewis acids onto urea scaffolds gives rise to a family of tunable hydrogen bond donor catalysts. The nature of the Lewis acid and associated ligands affects the urea polarization, acidity, and activity in reactions of nitrocyclopropane carboxylates and nitrodiazoesters.