MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Classification of epidermal keratins according to their immunoreactivity, isoelectric point, and mode of expression.
Classification of epidermal keratins according to their immunoreactivity, isoelectric point, and mode of expression.
The Journal of cell biology (1984-04-01)
R Eichner, P Bonitz, T T Sun
ABSTRACT
Human epidermal keratinocytes express under various growth conditions a total of at least nine keratins that can be divided into two subfamilies. Subfamily A comprises 40-, 46-, 48-, 50-/50'-, and 56.5-kilodalton (kd) keratins which are relatively acidic (pI less than 5.5) and, with the exception of 46-kd keratin, are recognized by AE1 monoclonal antibody. Subfamily B comprises 52-, 56-, 58-, and 65-67-kd keratins which are relatively basic (pI greater than 6) and are recognized by AE3 monoclonal antibody. Within each keratin subfamily, there is a constant member (50-/50'- and 58-kd keratins of the subfamilies A and B, respectively) that is always expressed. The other seven keratins of both subfamilies are variable members whose expression depends upon the cellular differentiated state, which is in turn modulated by the growth environment. The 56.5-kd keratin (subfamily A) and the 65-67-kd keratins (subfamily B) are coordinately expressed during keratinization. In contrast, the 40-, 46-, and 48-kd keratins (subfamily A) and the 52- and 56-kd keratins (subfamily B) are characteristic of cultured epidermal cells forming nonkeratinized colonies. These results demonstrate that human epidermal keratins can be classified according to their reactivity with monoclonal antikeratin antibodies, isoelectric point, and mode of expression. The classification of keratins into various subgroups may have important implications for the mechanisms of epidermal differentiation, the evolution of keratin heterogeneity, and the use of keratin markers for tumor diagnosis.
Sigma-Aldrich
Cytokeratin (35betaH11) Mouse Monoclonal Antibody
Sigma-Aldrich
Cytokeratin (OSCAR) Mouse Monoclonal Antibody