MilliporeSigma
  • Histopathological analyses of silent pituitary somatotroph adenomas using immunohistochemistry, in situ hybridization and confocal laser scanning microscopic observation.
Histopathological analyses of silent pituitary somatotroph adenomas using immunohistochemistry, in situ hybridization and confocal laser scanning microscopic observation.
Pathology, research and practice (2001-02-24)
A Matsuno, J Itoh, Y Itoh, R Y Osamura, H Katayama, T Nagashima
ABSTRACT
To characterize the morphological and functional aspects of silent somatotroph adenomas with paradoxical responses of GH in TRH or GnRH provocation tests, which are considered to be a useful strategy for endocrinological identification of silent somatotroph adenomas, we examined three silent somatotroph adenomas histopathologically. The adenomas were investigated by immunohistochemistry, including the highly sensitive catalyzed signal amplification system, the non-radioisotopic in situ hybridization method, and confocal laser scanning microscopy. GH production and GH-immunopositive secretory granules in the adenoma cells were demonstrated histopathologically, and the adenomas were interpreted as being densely granulated somatotroph adenomas. Endocrinological identification of silent somatotroph adenomas in combination with paradoxical responses of GH in TRH or GnRH provocation tests may elucidate the increasing number of silent somatotroph adenomas that have been regarded as mammotroph or clinically inactive adenomas. One should be aware of the differences between the previously reported silent somatotroph adenomas, most of which are sparsely granulated somatotroph adenomas, a somatotroph adenomas with paradoxical and the silent somatotroph adenomas, most of which are sparsely granulated somatotroph adenomas, and the silent somatotroph adenomas with paradoxical responses of GH in TRH or GnRH provocation tests, which are densely granulated somatotroph adenomas.
Sigma-Aldrich
GH (EP267) Rabbit Monoclonal Primary Antibody
Sigma-Aldrich
GH Rabbit Polyclonal Antibody