MilliporeSigma
  • Synthesis of colloidal silica nanoparticles of a tunable mesopore size and their application to the adsorption of biomolecules.
Synthesis of colloidal silica nanoparticles of a tunable mesopore size and their application to the adsorption of biomolecules.
Journal of colloid and interface science (2010-06-24)
Antonio B Fuertes, Patricia Valle-Vigón, Marta Sevilla
ABSTRACT
A synthetic method for the fabrication of colloidal mesoporous silica nanoparticles of a tunable mesopore size is presented. These nanoparticles form stable colloidal suspensions, have narrow particle size distributions in the 50-300nm range and exhibit a framework-confined porosity made up of large mesopores with a diameter of up to 9nm. The size of the mesopores was enlarged by means of two different swelling agents (SA), i.e. 1,3,5-trimethylbenzene (TMB) and N,N-dimethylhexadecylamine (DMHA). Colloidal mesoporous silica nanoparticles exhibit a strong affinity for the adsorption of hemoglobin (Hb) and the immobilization of lysozyme (Lz). These nanoparticles can adsorb up to 2080mgHbg(-1) support and 690mgLzg(-1) support. Moreover, the adsorption of these biomolecules on silica nanoparticles occurs very quickly and most of what is adsorbed is accommodated on the external surface area of the silica nanoparticles.
Sigma-Aldrich
N,N-Dimethylhexadecylamine, technical, ≥95% (GC)