MilliporeSigma
  • Clinical significance and biological roles of CRKL in human bladder carcinoma.
Clinical significance and biological roles of CRKL in human bladder carcinoma.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2014-01-01)
Bin Han, Lan Luan, Zhenqun Xu, Bin Wu
ABSTRACT
CRKL encodes an adaptor protein that has been recently reported to be overexpressed in various cancers and associate with the malignant behavior of cancer cells. However, the expression pattern of CRKL protein and its clinical significance in human bladder cancer have not been well characterized to date. In the present study, CRKL expression was analyzed in 82 archived bladder cancer specimens using immunohistochemistry, and the correlations between CRKL expression and clinicopathological parameters were evaluated. We found that CRKL was overexpressed in 31 of 82 (37.8%) bladder cancer specimens. A significant association was observed between CRKL overexpression and tumor status (p = 0.019). To further explore the biological functions of CRKL in bladder cancer, we overexpressed CRKL in BIU-87 and 5637 cell lines. Using CCK8 assay and colony formation assay, we showed that CRKL upregulation increased cell proliferation. In addition, transwell assay showed that CRKL could also facilitate invasion. Further study demonstrated that CRKL upregulation increased cyclin D1 expression and ERK phosphorylation. In conclusion, CRKL is overexpressed in bladder cancer and regulates malignant cell growth and invasion, which makes CRKL a candidate therapeutic target for bladder cancer.