MilliporeSigma
  • Monoclonal antibody capture enzyme-linked immunosorbent assay for detection of bovine enteric coronavirus.
Monoclonal antibody capture enzyme-linked immunosorbent assay for detection of bovine enteric coronavirus.
Journal of clinical microbiology (1984-03-01)
C F Crouch, T J Raybould, S D Acres
ABSTRACT
Monoclonal antibodies reactive with three different viral polypeptides were evaluated singly and in combination as the capture antibody(s) in an enzyme-linked immunosorbent assay system for the detection of bovine enteric coronavirus. Similar levels of sensitivity were found for all combinations tested. A sensitive, highly specific, and reproducible assay for the detection of bovine enteric coronavirus was developed, using a mixture of two of these monoclonal antibodies reactive with antigenic components either external or internal to the virion. These monoclonal antibodies were bound indirectly to 96-well plates via rabbit anti-mouse immunoglobulin. After sample application and incubation, virus was detected by using rabbit anti-coronavirus peroxidase conjugate followed by enzyme substrate and chromagen. Fecal samples from a single herd of cows were screened for the presence of coronavirus by this assay. Five percent of clinically normal cows were found to be shedding coronavirus.
Sigma-Aldrich
Poly-L-lysine hydrobromide, mol wt 70,000-150,000 by viscosity
Pricing and availability is not currently available.