MilliporeSigma
  • A need for determination of arsenic species at low levels in cereal-based food and infant cereals. Validation of a method by IC-ICPMS.
A need for determination of arsenic species at low levels in cereal-based food and infant cereals. Validation of a method by IC-ICPMS.
Food chemistry (2013-11-12)
Toni Llorente-Mirandes, Josep Calderón, Francesc Centrich, Roser Rubio, José Fermín López-Sánchez
ABSTRACT
The present study arose from the need to determine inorganic arsenic (iAs) at low levels in cereal-based food. Validated methods with a low limit of detection (LOD) are required to analyse these kinds of food. An analytical method for the determination of iAs, methylarsonic acid (MA) and dimethylarsinic acid (DMA) in cereal-based food and infant cereals is reported. The method was optimised and validated to achieve low LODs. Ion chromatography-inductively coupled plasma mass spectrometry (IC-ICPMS) was used for arsenic speciation. The main quality parameters were established. To expand the applicability of the method, different cereal products were analysed: bread, biscuits, breakfast cereals, wheat flour, corn snacks, pasta and infant cereals. The total and inorganic arsenic content of 29 cereal-based food samples ranged between 3.7-35.6 and 3.1-26.0 μg As kg(-1), respectively. The present method could be considered a valuable tool for assessing inorganic arsenic contents in cereal-based foods.
Product Description
Rice flour (Total As and As species), ERM®, certified reference material