MilliporeSigma
Anaphylaxis due to caffeine.
Asia Pacific allergy (2015-02-06)
Kumiya Sugiyama, Tatsurai Cho, Masamitsu Tatewaki, Shogo Onishi, Tatsuya Yokoyama, Naruo Yoshida, Takayoshi Fujimatsu, Hirokuni Hirata, Takeshi Fukuda, Yasutsugu Fukushima
ABSTRACT
We report a rare case of anaphylaxis due to caffeine intake. A 27-year-old woman suffered her first episode of anaphylaxis and a positive skin prick test suggested that the anaphylaxis was due to an IgE-mediated hypersensitivity reaction to caffeine. She was diagnosed with caffeine allergy and has not had an allergic reaction after avoiding foods and drinks containing caffeine. Although caffeine is known to have antiallergic effects, this case shows that caffeine can be an allergen and cause anaphylaxis.
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG