MilliporeSigma
  • Enantiomeric resolution of ibuprofen and flurbiprofen in human plasma by SPE-chiral HPLC methods.
Enantiomeric resolution of ibuprofen and flurbiprofen in human plasma by SPE-chiral HPLC methods.
Combinatorial chemistry & high throughput screening (2012-05-11)
Imran Ali, Iqbal Hussain, Kishwar Saleem, Hassan Y Aboul-Enein
ABSTRACT
Chiral analysis of profens in human plasma is an important area of research due to different pharmaceutical activities of their enantiomers. The solid phase extraction of ibuprofen and flurbiprofen from human plasma was carried out on C18 cartridges by using phosphate buffer (50 mM, pH 6.0) followed by elution with methanol. Chiral-HPLC was performed on AmyCoat RP (150 mm x 46 mm, 3 μm particle size) column by using different combinations of water-acetonitrile-trifluoro acetic acid at 1.5 mLmin-1 flow rate. The detection was achieved at 236 and 254 nm for ibuprofen and flurbiprofen, respectively with 27±1°C as working temperature. The chromatographic parameters i.e. retention (k), separation (α) and resolution (Rs) factors ranged from 4.54-14.42, 1.10-1.30 and 1.01-1.49, respectively. The binding differences of enantiomers of ibuprofen and flurbiprofen were 4.4 and 5.2, respectively. These values suggest that S-(+)- enantiomer of flurbiprofen is more active than ibuprofen due to low enantiomeric difference of the later drug. The developed SPE-Chiral HPLC methods were validated, which are selective, efficient and reproducible.