MilliporeSigma
  • Human laminin B2 chain. Comparison of the complete amino acid sequence with the B1 chain reveals variability in sequence homology between different structural domains.
Human laminin B2 chain. Comparison of the complete amino acid sequence with the B1 chain reveals variability in sequence homology between different structural domains.
The Journal of biological chemistry (1988-05-15)
T Pikkarainen, T Kallunki, K Tryggvason
ABSTRACT
The complete amino acid sequence of the human laminin B2 chain has been determined by sequencing of cDNA clones. The six overlapping clones studied cover approximately 7.5 kilobases of which 5312 nucleotides were sequenced from the 5' end. The open reading frame codes for a 33-residue signal peptide and a 1576-residue B2 chain proper, which is 189 residues less than in the highly homologous B1 chain (Pikkarainen, T., Eddy, R., Fukushima, Y., Byers, M., Shows, T., Pihlajaniemi, T., Saraste, M., and Tryggvason, K. (1987) J. Biol. Chem. 262, 10454-10462). Computer analysis revealed that the B2 chain consists of distinct domains that contain helical structures, cysteine-rich repeats, and globular regions, as does the B1 chain. However, domain alpha and domain beta of the B1 chain have no counterpart in B2, and the number of cysteine-rich repeats is 12, or 1 less than in the B1 chain. The degree of homology between the two chains is highest in the cysteine repeat-containing domains III and V where 40% of the residues match. However, results demonstrate that the B1 and B2 chains of laminin are highly homologous proteins that are probably the products of related genes.