MilliporeSigma
  • Bid-induced mitochondrial membrane permeabilization waves propagated by local reactive oxygen species (ROS) signaling.
Bid-induced mitochondrial membrane permeabilization waves propagated by local reactive oxygen species (ROS) signaling.
Proceedings of the National Academy of Sciences of the United States of America (2012-03-07)
Cecilia Garcia-Perez, Soumya Sinha Roy, Shamim Naghdi, Xuena Lin, Erika Davies, György Hajnóczky
ABSTRACT
Bid-induced mitochondrial membrane permeabilization and cytochrome c release are central to apoptosis. It remains a mystery how tiny amounts of Bid synchronize the function of a large number of discrete organelles, particularly in mitochondria-rich cells. Looking at cell populations, the rate and lag time of the Bid-induced permeabilization are dose-dependent, but even very low doses lead eventually to complete cytochrome c release. By contrast, individual mitochondria display relatively rapid and uniform kinetics, indicating that the dose dependence seen in populations is due to a spreading of individual events in time. We report that Bid-induced permeabilization and cytochrome c release regularly demonstrate a wave-like pattern, propagating through a cell at a constant velocity without dissipation. Such waves do not depend on caspase activation or permeability transition pore opening. However, reactive oxygen species (ROS) scavengers suppressed the coordination of cytochrome c release and also inhibited Bid-induced cell death, whereas both superoxide and hydrogen peroxide sensitized mitochondria to Bid-induced permeabilization. Thus, Bid engages a ROS-dependent, local intermitochondrial potentiation mechanism that amplifies the apoptotic signal as a wave.