MilliporeSigma
  • Human DEAD-box ATPase DDX3 shows a relaxed nucleoside substrate specificity.
Human DEAD-box ATPase DDX3 shows a relaxed nucleoside substrate specificity.
Proteins (2007-03-16)
Raffaella Franca, Amalia Belfiore, Silvio Spadari, Giovanni Maga
ABSTRACT
Human DDX3 (hDDX3) is a DEAD-box protein shown to possess RNA-unwinding and adenosine triphosphatase (ATPase) activities. The hDDX3 protein has been implicated in nuclear mRNA export, cell growth control, and cancer progression. In addition, a role of this protein in the replication of human immunodeficiency virus Type 1 and in the pathogenesis of hepatitis C virus has been recently proposed. Its enzymological properties, however, are largely unknown. In this work, we characterized its ATPase activity. We show that hDDX3 ATPase activity is stimulated by various ribo- and deoxynucleic acids. Comparative analysis with different nucleoside triphosphate analogs showed that the hDDX3 ATPase couples high catalytic efficiency to a rather relaxed substrate specificity, both in terms of base selection and sugar selection. In addition, its ability to recognize the L-stereoisomers of both 3' deoxy- and 2',3' dideoxy-ribose, points to a relaxed stereoselectivity. On the basis of these results, we hypothesize the presence of structural determinants on both the base and the sugar moieties, critical for nucleoside binding to the enzyme. Our results expand the knowledge about the DEAD-box RNA helicases in general and can be used for rational design of selective inhibitors of hDDX3, to be tested as potential antitumor and antiviral agents.