MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Efficient synthesis of a chiral precursor for angiotensin-converting enzyme (ACE) inhibitors in high space-time yield by a new reductase without external cofactors.
Efficient synthesis of a chiral precursor for angiotensin-converting enzyme (ACE) inhibitors in high space-time yield by a new reductase without external cofactors.
Organic letters (2012-04-07)
Nai-Dong Shen, Yan Ni, Hong-Min Ma, Li-Juan Wang, Chun-Xiu Li, Gao-Wei Zheng, Jie Zhang, Jian-He Xu
ABSTRACT
A new reductase, CgKR2, with the ability to reduce ethyl 2-oxo-4-phenylbutyrate (OPBE) to ethyl (R)-2-hydroxy-4-phenylbutyrate ((R)-HPBE), an important chiral precursor for angiotensin-converting enzyme (ACE) inhibitors, was discovered. For the first time, (R)-HPBE with >99% ee was produced via bioreduction of OPBE at 1 M without external addition of cofactors. The space-time yield (700 g·L(-1)·d(-1)) was 27 times higher than the highest record.