MilliporeSigma
  • Stable plasma-deposited acrylic acid surfaces for cell culture applications.
Stable plasma-deposited acrylic acid surfaces for cell culture applications.
Biomaterials (2005-01-01)
Loredana Detomaso, Roberto Gristina, Giorgio S Senesi, Riccardo d'Agostino, Pietro Favia
ABSTRACT
Continuous and modulated glow discharges were used to deposit thin films from acrylic acid vapors. Different deposition regimes were investigated, and their effect on chemical composition, morphology and homogeneity of the coatings, as well as on their stability in water and resistance to sterilization. Stable films were utilized in cell adhesion experiments with human fibroblasts.
Sigma-Aldrich
Acrylic acid, anhydrous, contains 200 ppm MEHQ as inhibitor, 99%