MilliporeSigma
  • Synthesis of Pt nanoparticles decorated 1,5-diaminoanthraquinone nanofibers and their application toward catalytic reduction of 4-nitrophenol.
Synthesis of Pt nanoparticles decorated 1,5-diaminoanthraquinone nanofibers and their application toward catalytic reduction of 4-nitrophenol.
Journal of nanoscience and nanotechnology (2012-10-06)
Guohui Chang, Yonglan Luo, Xiaoyun Qin, Wenbo Lu, Abdullah M Asiri, Abdulrahman O Al-Youbi, Xuping Sun
ABSTRACT
The present communication reports on the rapid preparation of 1,5-diaminoanthraquinone nanofibers (DAAQNFs) on a large scale by a reprecipitation method and their subsequent decoration with small platinum nanoparticles (PtNPs) using tannic acid (TA) as a reducing agent. It suggests the resultant PtNP/DAAQNF composites exhibit a good catalytic activity toward reduction of 4-nitrophenol (4-NP) to 4-aminophenol (4-AP) by NaBH4. It also suggests that the composites exhibit higher catalytic activity than the PtNPs due to that the DAAQNF support may play an active part in the catalysis, yielding a synergistic effect. The possible mechanism involved has also been discussed.
Sigma-Aldrich
1,5-Diaminoanthraquinone, technical grade, 85%
Pricing and availability is not currently available.