MilliporeSigma
  • Phosphorylation and sulfation of oligosaccharide substrates critically influence the activity of human beta1,4-galactosyltransferase 7 (GalT-I) and beta1,3-glucuronosyltransferase I (GlcAT-I) involved in the biosynthesis of the glycosaminoglycan-protein linkage region of proteoglycans.
Phosphorylation and sulfation of oligosaccharide substrates critically influence the activity of human beta1,4-galactosyltransferase 7 (GalT-I) and beta1,3-glucuronosyltransferase I (GlcAT-I) involved in the biosynthesis of the glycosaminoglycan-protein linkage region of proteoglycans.
The Journal of biological chemistry (2004-11-04)
Sandrine Gulberti, Virginie Lattard, Magali Fondeur, Jean-Claude Jacquinet, Guillermo Mulliert, Patrick Netter, Jacques Magdalou, Mohamed Ouzzine, Sylvie Fournel-Gigleux
ABSTRACT
We determined whether the two major structural modifications, i.e. phosphorylation and sulfation of the glycosaminoglycan-protein linkage region (GlcAbeta1-3Galbeta1-3Galbeta1-4Xylbeta1), govern the specificity of the glycosyltransferases responsible for the biosynthesis of the tetrasaccharide primer. We analyzed the influence of C-2 phosphorylation of Xyl residue on human beta1,4-galactosyltransferase 7 (GalT-I), which catalyzes the transfer of Gal onto Xyl, and we evaluated the consequences of C-4/C-6 sulfation of Galbeta1-3Gal (Gal2-Gal1) on the activity and specificity of beta1,3-glucuronosyltransferase I (GlcAT-I) responsible for the completion of the glycosaminoglycan primer sequence. For this purpose, a series of phosphorylated xylosides and sulfated C-4 and C-6 analogs of Galbeta1-3Gal was synthesized and tested as potential substrates for the recombinant enzymes. Our results revealed that the phosphorylation of Xyl on the C-2 position prevents GalT-I activity, suggesting that this modification may occur once Gal is attached to the Xyl residue of the nascent oligosaccharide linkage. On the other hand, we showed that sulfation on C-6 position of Gal1 of the Galbeta1-3Gal analog markedly enhanced GlcAT-I catalytic efficiency and we demonstrated the importance of Trp243 and Lys317 residues of Gal1 binding site for enzyme activity. In contrast, we found that GlcAT-I was unable to use digalactosides as acceptor substrates when Gal1 was sulfated on C-4 position or when Gal2 was sulfated on both C-4 and C-6 positions. Altogether, we demonstrated that oligosaccharide modifications of the linkage region control the specificity of the glycosyltransferases, a process that may regulate maturation and processing of glycosaminoglycan chains.
Sigma-Aldrich
7-Methoxy-2-naphthol, 98%