MilliporeSigma
  • Diastereoselective formal synthesis of a monoterpene alkaloid, (-)-incarvilline.
Diastereoselective formal synthesis of a monoterpene alkaloid, (-)-incarvilline.
The Journal of organic chemistry (2007-07-31)
Toshio Honda, Kyosuke Kaneda
ABSTRACT
Diastereoselective formal synthesis of a monoterpene alkaloid, (-)-incarvilline, the key intermediate for the synthesis of (-)-incarvillateine, was achieved by using an intramolecular Pauson-Khand reaction of (S)-N-[(E)-2-butenyl]-N-(3-butynyl-2-methoxymethoxy)-p-toluenesulfonamide as a key step.
Sigma-Aldrich
N-(tert-Butoxycarbonyl)-p-toluenesulfonamide
Pricing and availability is not currently available.