MilliporeSigma
  • Microwave-assisted solution- and solid-phase synthesis of 2-amino-4-arylpyrimidine derivatives.
Microwave-assisted solution- and solid-phase synthesis of 2-amino-4-arylpyrimidine derivatives.
Journal of combinatorial chemistry (2007-03-14)
Mitra Matloobi, C Oliver Kappe
ABSTRACT
An efficient and rapid microwave-assisted solution-phase method for the synthesis of 2-amino-4-arylpyrimidine-5-carboxylic acid derivatives has been developed. The five-step linear protocol involves an initial Biginelli multicomponent reaction leading to dihydropyrimidine-2-thiones which are subsequently S-alkylated with methyl iodide. The resulting 2-methylthiodihydropyrimidines are sequentially oxidized first with manganese dioxide and then with Oxone to provide 2-methylsulfonyl-pyrimidines which serve as excellent precursors for the generation of a variety of 2-substituted pyrimidines via displacement of the reactive sulfonyl group with nitrogen, oxygen, sulfur, and carbon nucleophiles. A modified protocol using a solid-phase method has also been developed.