MilliporeSigma
  • Expanding the chemical diversity of spirooxindoles via alkylative pyridine dearomatization.
Expanding the chemical diversity of spirooxindoles via alkylative pyridine dearomatization.
Beilstein journal of organic chemistry (2012-09-29)
Chunhui Dai, Bo Liang, Corey R J Stephenson
ABSTRACT
A mild and practical synthesis of spirooxindole [1,3]oxazino derivatives from N-substituted isatins and 1,3-dicarbonyl compounds with pyridine derivatives is reported. The reactions provided good to excellent yields. Further exploration of the molecular diversity of these compounds is demonstrated through Diels-Alder reactions.