MilliporeSigma
  • Synthesis of a tetrasaccharide glycosyl glycerol. Precursor to glycolipids of Meiothermus taiwanensis ATCC BAA-400.
Synthesis of a tetrasaccharide glycosyl glycerol. Precursor to glycolipids of Meiothermus taiwanensis ATCC BAA-400.
The Journal of organic chemistry (2007-06-21)
Chien-Tai Ren, Yu-Hsuan Tsai, Yu-Liang Yang, Wei Zou, Shih-Hsiung Wu
ABSTRACT
Synthesis of a tetrasaccharide glycosyl glycerol, the core structure of glycoglycerolipid from Meiothermus taiwanensis ATCC BAA-400, was described. A one-pot glycosylation with three components was employed as a key step.