MilliporeSigma
  • Nuclear-localized subtype of end-binding 1 protein regulates spindle organization in Arabidopsis.
Nuclear-localized subtype of end-binding 1 protein regulates spindle organization in Arabidopsis.
Journal of cell science (2010-01-14)
Shinichiro Komaki, Tatsuya Abe, Silvie Coutuer, Dirk Inzé, Eugenia Russinova, Takashi Hashimoto
ABSTRACT
End-binding 1 (EB1) proteins are evolutionarily conserved plus-end-tracking proteins that localize to growing microtubule plus ends where they regulate microtubule dynamics and interactions with intracellular targets. Animal EB1 proteins have acidic C-terminal tails that might induce an autoinhibitory conformation. Although EB1 proteins with the same structural features occur in plants (EB1a and EB1b in Arabidopsis thaliana), a variant form (EB1c) is present that lacks the characteristic tail. We show that in Arabidopsis the tail region of EB1b, but not of EB1c, inhibits microtubule assembly in vitro. EB1a and EB1b form heterodimers with each other, but not with EB1c. Furthermore, the EB1 genes are expressed in various cell types of Arabidopsis, but the expression of EB1c is particularly strong in the meristematic cells where it is targeted to the nucleus by a nuclear localization signal in the C-terminal tail. Reduced expression of EB1c compromised the alignment of spindle and phragmoplast microtubules and caused frequent lagging of separating chromosomes at anaphase. Roots of the eb1c mutant were hypersensitive to a microtubule-disrupting drug and complete rescue of the mutant phenotype required the tail region of EB1c. These results suggest that a plant-specific EB1 subtype has evolved to function preferentially on the spindle microtubules by accumulating in the prophase nucleus.