MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Asymmetric synthesis of a prostaglandin D2 receptor antagonist.
The Journal of organic chemistry (2004-12-31)
Kevin R Campos, Michel Journet, Sandra Lee, Edward J J Grabowski, Richard D Tillyer
ABSTRACT
An asymmetric synthesis was developed for the production of a prostaglandin D(2) receptor antagonist for the treatment of allergic rhinitis. The stereogenic center was set using asymmetric allylic alkylation chemistry, and the core of the structure was constructed via Pd-catalyzed N-cyclization/Heck methodology. The synthesis relies on a late stage indoline oxidation which does not racemize the product.