MilliporeSigma
  • Dicarba-closo-dodecaboranes as a pharmacophore. Retinoidal antagonists and potential agonists.
Dicarba-closo-dodecaboranes as a pharmacophore. Retinoidal antagonists and potential agonists.
Chemical & pharmaceutical bulletin (1999-04-23)
T Iijima, Y Endo, M Tsuji, E Kawachi, H Kagechika, K Shudo
ABSTRACT
Synthesis and biological evaluation of the first dicarba-closo-dodecaborane (carborane) derivatives of retinoids are described. Their retinoidal activity were examined in terms of the differentiation-inducing ability toward human promyelocytic leukemia HL-60 cells. High retinoidal activity (agonist or antagonist for retinoic acid receptor (RAR) requires a carboxylic acid moiety and an appropriate hydrophobic group located at a suitable position on the molecule. The 4-carboranyl-substituted compounds (7, 11) showed antagonistic activity but no agonistic activity even in the presence of the potent synergist HX630. On the other hand, the 3-carboranyl-substituted compounds (8, 12) showed potential agonistic activity, but no antagonistic activity. The results indicates that carboranes are applicable as the hydrophobic moiety of biologically active molecules.
Sigma-Aldrich
2-Ethynylanisole, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethyl 3-bromobenzoate, 98%
Pricing and availability is not currently available.