MilliporeSigma
  • Determination of the absolute configuration of the active amlodipine enantiomer as (-)-S: a correction.
Determination of the absolute configuration of the active amlodipine enantiomer as (-)-S: a correction.
Journal of medicinal chemistry (1992-09-04)
S Goldmann, J Stoltefuss, L Born
ABSTRACT
The active (-) enantiomer of amlodipine was originally reported to have R configuration. This does not concur with other 1,4-dihydropyridines with known absolute configuration. This configuration has now been determined by X-ray structural analysis using (1S)-camphanic acid and (S)-2-methoxy-2-phenylethanol as chiral probes. Both determinations gave the S configuration for the amlodipine (-) enantiomer with the greater Ca-antagonistic activity.