MilliporeSigma
  • Overexpression of human prostaglandin G/H synthase-1 and -2 by recombinant vaccinia virus: inhibition by nonsteroidal anti-inflammatory drugs and biosynthesis of 15-hydroxyeicosatetraenoic acid.
Overexpression of human prostaglandin G/H synthase-1 and -2 by recombinant vaccinia virus: inhibition by nonsteroidal anti-inflammatory drugs and biosynthesis of 15-hydroxyeicosatetraenoic acid.
Molecular pharmacology (1994-02-01)
G P O'Neill, J A Mancini, S Kargman, J Yergey, M Y Kwan, J P Falgueyret, M Abramovitz, B P Kennedy, M Ouellet, W Cromlish
ABSTRACT
Human prostaglandin G/H synthase (hPGHS)-1 and hPGHS-2, key enzymes in the formation of prostanoids from arachidonic acid, were expressed at high levels in COS-7 cells using a T7 RNA polymerase/vaccinia virus expression system. The open reading frame of hPGHS-2 cloned into vaccinia virus without its natural 5' and 3' untranslated regions directed only low levels of hPGHS-2 enzyme activity in COS-7 cells. High-level hPGHS-2 expression was achieved by appending the 3' untranslated region of hPGHS-1 to the hPGHS-2 open reading frame, with subsequent expression of the hybrid mRNA using vaccinia virus. Enzymatically active recombinant hPGHS-1 and hPGHS-2 were present as glycosylated proteins in the microsomal fraction prepared from infected cells, whereas recombinant hPGHS-1 and hPGHS-2 prepared from the microsomal fraction of cells treated with tunicamycin, an inhibitor of N-linked glycosylation, were enzymatically inactive. The major prostanoid products formed by microsomes from COS-7 cells containing either recombinant hPGHS-1 or hPGHS-2 after incubation with arachidonic acid were prostaglandin D2 and E2, with lower levels of prostaglandin F2 alpha and 6-keto-prostaglandin F1 alpha. A range of potencies were observed for various nonsteroidal anti-inflammatory drugs as inhibitors of prostaglandin E2 synthesis by hPGHS-1 and hPGHS-2. Recombinant hPGHS-1 and hPGHS-2 both produced 15- and 11-hydroxyeicosatetraenoic acid (HETE) from arachidonic acid, with 15-HETE production by hPGHS-2 being stimulated 5-fold by preincubation with aspirin. Chiral phase high performance liquid chromatography analysis showed that aspirin-treated hPGHS-2 produced 15(R)-HETE, with no detectable 15(S)-HETE.