MilliporeSigma
  • Thermodynamic study of sesamol, piperonyl alcohol, piperonylic acid and homopiperonylic acid: a combined experimental and theoretical investigation.
Thermodynamic study of sesamol, piperonyl alcohol, piperonylic acid and homopiperonylic acid: a combined experimental and theoretical investigation.
Organic & biomolecular chemistry (2004-03-10)
M Agostinha R Matos, Manuel J S Monte, Clara C S Sousa, Ana R R P Almeida, Victor M F Morais
ABSTRACT
The standard (p(o)= 0.1 MPa) molar energies of combustion in oxygen, at T= 298.15 K, of four 1,3-benzodioxole derivatives (sesamol, piperonyl alcohol, piperonylic acid and homopiperonylic acid) were measured by static bomb calorimetry. The values of the standard molar enthalpies of sublimation, at T= 298.15 K, were derived from vapour pressure-temperature measurements using the Knudsen effusion technique. Combining these results the standard molar enthalpies of formation of the compounds, in the gas phase, at T= 298.15 K, have been calculated: sesamol (-325.7 +/- 1.9) kJ mol(-1); piperonyl alcohol (-329.0 +/- 2.0) kJ mol(-1); piperonylic acid (-528.9 +/- 2.6) kJ mol(-1) and homopiperonylic acid (-544.5 +/- 2.9) kJ mol(-1). The most stable geometries of all the compounds were obtained using the density functional theory with the B3LYP functional and two basis sets: 6-31G** and 6-311G**. The nonplanarity of the molecules was analyzed in terms of the anomeric effect, which is believed to arise from the interaction between a nonbonded oxygen p orbital and the empty orbital sigma*(CO) involving the other oxygen atom. Calculations were performed to obtain estimates of the enthalpies of formation of all the benzodioxoles using appropriate isodesmic reactions. There is a perfect agreement between theoretical and experimental results.
Sigma-Aldrich
3,4-(Methylenedioxy)phenylacetic acid, 98%
Pricing and availability is not currently available.