MilliporeSigma
  • Identification of 2-[2-nitro-4-(trifluoromethyl)benzoyl]- cyclohexane-1,3-dione metabolites in urine of patients suffering from tyrosinemia type I with the use of 1H and 19F NMR spectroscopy.
Identification of 2-[2-nitro-4-(trifluoromethyl)benzoyl]- cyclohexane-1,3-dione metabolites in urine of patients suffering from tyrosinemia type I with the use of 1H and 19F NMR spectroscopy.
Acta biochimica Polonica (2008-11-29)
Przemysław Szczeciński, Diana Lamparska, Adam Gryff-Keller, Wanda Gradowska
ABSTRACT
Organic extracts of six urine samples from children treated with nitisinone, a medicine against tyrosinemia type I, were investigated by (1)H and (19)F NMR spectroscopy. The presence of unchanged 2-[2-nitro-4-(trifluoromethyl)benzoyl]cyclohexane-1,3-dione (NTBC), 6-hydroxy-2-[2-nitro-4-(trifluoromethyl)benzoyl]cyclohexane-1,3-dione (NTBC-OH) and 2-nitro-4-trifluoromethylbenzoic acid (NTFA) as well as a few other unidentified compounds containing CF(3) group was documented.
Sigma-Aldrich
2-Nitro-4-(trifluoromethyl)benzoic acid, 98%
Pricing and availability is not currently available.