MilliporeSigma
  • In situ derivatization hollow fiber mediated liquid phase microextraction of alkylphosphonic acids from water.
In situ derivatization hollow fiber mediated liquid phase microextraction of alkylphosphonic acids from water.
Journal of chromatography. A (2006-12-19)
Deepak Pardasani, Pankaj K Kanaujia, A K Gupta, Vijay Tak, R K Shrivastava, D K Dubey
ABSTRACT
Alkylphosphonic acids (APAs), particularly the methyl-, ethyl-, isopropyl- and n-propyl-phosphonic acids are important markers of extremely toxic nerve agents. Hence, their detection and identification is of vital importance to verification of chemical weapons convention (CWC). Verification analysis of CWC requires development of fast, reliable, simple and reproducible sample preparation methods of water and soil samples. Present investigation is focused on the optimization of alkylation of APAs in water with subsequent extraction of alkylated acids by hollow fiber liquid phase microextraction (HF-LPME). This simple and sensitive sample preparation of APAs from water offered better recoveries in comparison to conventionally used extraction technique. Under optimized conditions, the APAs were detected at the concentration of 0.5-0.75 microg/mL with S/N ratio > or = 5, whereas the LODs for alkyl APAs (monobasic APAs) were achieved up to 0.1 microg/mL. The developed method was finally tested with water samples supplied in 19th official proficiency test conducted by the OPCW.
Sigma-Aldrich
Propylphosphonic acid, 95%