- Efficient regioselective synthesis and potential antitumor evaluation of isoxazolo[5,4-b]pyridines and related annulated compounds.
Efficient regioselective synthesis and potential antitumor evaluation of isoxazolo[5,4-b]pyridines and related annulated compounds.
The reaction of 5-amino-3-methylisoxazole with appropriate α,β-unsaturated ketones gave the corresponding isoxazolo[5,4-b]pyridines. Treatment of 1 with 2,6-dibenzylidenecyclohexanone or 2-benzylidenedimedone afforded the corresponding isoxazolo[5,4-b]quinoline derivatives. 4,6,8,9-Tetrahydroisoxazolo[5,4-b]quinolin-5-one derivative was also obtained by multicomponent condensation reaction of 1 with dimedone and benzaldehyde. Heterocyclic annulation of the isoxazolo[5,4-b]pyridine system was achieved via reaction of 1 with benzylidene derivatives of indandione, quinuclidone, pyrazolone, and oxazolone. A representative of some newly synthesized compounds was evaluated as antitumor agents.