MilliporeSigma
  • The synthesis and characterization of nonperipherally tetra terminal alkynyl substituted phthalocyanines and glycoconjugation via the click reaction.
The synthesis and characterization of nonperipherally tetra terminal alkynyl substituted phthalocyanines and glycoconjugation via the click reaction.
Dalton transactions (Cambridge, England : 2003) (2014-04-26)
Zeliha Kanat, Hatice Dinçer
ABSTRACT
In order to obtain nonperipherally tetra terminal alkynyl substituted phthalocyanines (Pcs), new 3-pent-4-ynyloxy phthalonitrile (3) was prepared by the nucleophilic displacement reaction of 3-nitrophthalonitrile (1) and 4-pentyn-1-ol (2) and then cyclotetramerization was attained in the presence of zinc acetate, cobalt acetate, and/or DBU in n-pentanol without protection/deprotection. For the first time, the glycoconjugation of the nonperipherally tetra terminal alkynyl substituted zinc phthalocyanine (ZnPc) (6) can be easily achieved via the click reaction in a high yield. The electronic absorption spectrum of the glucopyranosyl substituted ZnPc (10) derivative showed a red-shifted Q band at 751 nm in dichloromethane due to the protonation of the meso nitrogens of the Pc macrocycle. Deacylation yielded ZnPc (11) bearing glucose substituents at nonperipheral positions with an improved water-solubility and non-aggregation in DMSO. The chemical structures of the new compounds were characterized by (1)H NMR, (13)C NMR, FT-IR, UV-Vis, mass spectrometry and elemental analysis. Moreover, the phthalonitrile compound was characterized using X-ray.
Sigma-Aldrich
4-Pentyn-1-ol, 97%