MilliporeSigma
  • Cycloruthenated complexes from imine-based heterocycles: synthesis, characterization, and reactivity toward alkynes.
Cycloruthenated complexes from imine-based heterocycles: synthesis, characterization, and reactivity toward alkynes.
Chemistry (Weinheim an der Bergstrasse, Germany) (2012-09-29)
Luciano Cuesta, Tatiana Soler, Esteban P Urriolabeitia
ABSTRACT
Novel cycloruthenated complexes 2 a-c, 4 a-c, and 6 a, b based on heteroaromatic cores have been synthesized by reaction of a series of heterocycle-based imines with [{RuCl(η(6)-p-cymene)}(2)(μ-Cl)(2)] and Cu(OAc)(2). This approach has proved efficient for the cyclometalation of thiophene, benzothiophene, furan, benzofuran, pyrrole, and indole derivatives. In addition, even a double cyclometalation process over the same heterocyclic ring is possible, yielding unprecedented bimetallic complexes. These ruthenacycles react with 3-hexyne through an unexpected pathway, which involves the coupling of the original imino heterocycle and acetylene followed by dearomatization to afford fused hetero-hydropyridyl ligands bonded to the {Ru(p-cymene)} organometallic moiety (i.e., 7 a-c and 8 a-c). These complexes represent, as far as we know, the first examples of this type of compound within the context of cyclometalation, and an exhaustive analysis of their structure was carried out in solution and solid state. Furthermore, these unique species react with CuCl(2), which promotes the rearomatization and the release of highly valuable aromatic fused bis-heterocycles (i.e., 9 a-c, 10 a-c, 11 a, and 12 a/12 a'), providing a novel and appealing synthetic route to this extraordinary family of molecules.
Sigma-Aldrich
3-Hexyne, 99%