MilliporeSigma
Fragrance material review on 3,5,5-trimethyl-1-hexanol.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2010-08-13)
D McGinty, J Scognamiglio, C S Letizia, A M Api
ABSTRACT
A toxicologic and dermatologic review of 3,5,5-trimethyl-1-hexanol when used as a fragrance ingredient is presented. 3,5,5-Trimethyl-1-hexanol is a member of the fragrance structural group branched chain saturated alcohols. The common characteristic structural elements of the alcohols with saturated branched chain are one hydroxyl group per molecule, and a C(4) to C(12) carbon chain with one or several methyl side chains. This review contains a detailed summary of all available toxicology and dermatology papers that are related to this individual fragrance ingredient and is not intended as a stand-alone document. A safety assessment of the entire branched chain saturated alcohol group will be published simultaneously with this document; please refer to Belsito et al. (2010) for an overall assessment of the safe use of this material and all other branched chain saturated alcohols in fragrances.
Sigma-Aldrich
3,5,5-Trimethyl-1-hexanol, technical grade, 85%