MilliporeSigma
  • Experimental and modeling study of the thermal decomposition of methyl decanoate.
Experimental and modeling study of the thermal decomposition of methyl decanoate.
Combustion and flame (2011-07-01)
Olivier Herbinet, Pierre-Alexandre Glaude, Valérie Warth, Frédérique Battin-Leclerc
ABSTRACT
The experimental study of the thermal decomposition of methyl decanoate was performed in a jet-stirred reactor at temperatures ranging from 773 to 1123 K, at residence times between 1 and 4 s, at a pressure of 800 Torr (106.6 kPa) and at high dilution in helium (fuel inlet mole fraction of 0.0218). Species leaving the reactor were analyzed by gas chromatography. Main reaction products were hydrogen, carbon oxides, small hydrocarbons from C
Sigma-Aldrich
Methyl decanoate, 99%