MilliporeSigma
Signaling mechanistics: aluminum fluoride for molecule of the year.
Current biology : CB (1998-02-28)
A Wittinghofer
ABSTRACT
Recent three-dimensional structures of phosphoryl transfer enzymes in their aluminum fluoride bound state and corresponding biochemical data have shown how diverse biological problems can be investigated using this small inorganic molecule.
Sigma-Aldrich
Aluminum fluoride, anhydrous, powder, 99.8% trace metals basis