MilliporeSigma
Recent progress in biocatalysis using supercritical carbon dioxide.
Journal of bioscience and bioengineering (2012-11-21)
Tomoko Matsuda
ABSTRACT
The latest advances in biocatalysis using supercritical carbon dioxide (scCO(2)) are reviewed. Stability and stabilization methodologies of enzymes in scCO(2) as well as reactions for organic synthesis are described. Especially, varieties of examples for lipase-catalyzed synthesis of chiral compounds using scCO(2) are given. Furthermore, asymmetric reduction by alcohol dehydrogenase in scCO(2) and carboxylation by decarboxylase in scCO(2) are also introduced.
Sigma-Aldrich
Carbon dioxide, ≥99.8%
Sigma-Aldrich
Carbon-12C dioxide, 99.9 atom % 12C
Sigma-Aldrich
Carbon-12C dioxide, 99.99 atom % 12C