MilliporeSigma
  • Why dapsone stops seizures and may stop neutrophils' delivery of VEGF to glioblastoma.
Why dapsone stops seizures and may stop neutrophils' delivery of VEGF to glioblastoma.
British journal of neurosurgery (2012-05-04)
R E Kast, F Lefranc, G Karpel-Massler, M-E Halatsch
ABSTRACT
Lopez-Gomez et al. recently published remarkable but mechanistically unexplained empirical evidence that the old antibiotic dapsone has antiepileptic activity. We addressed the question "Why should a sulfone antibiotic reduce seizures?". We report here our conclusions based on data from past studies that seizures are associated with elevated interleukin-8 (IL-8) and that dapsone inhibits IL-8 release and function in several different clinical and experimental contexts. Diverse CNS insults cause an increase in CNS IL-8. Thus, the pro-inflammatory environment generated by increase IL-8 leads to a lower seizure threshold. Together this evidence indicates dapsone exerts anti-seizure activity by diminishing IL-8 signalling. Since IL-8 is clearly upregulated in glioblastoma and contributes to the florid angiogenesis of that disease, and since interference with IL-8 function has been shown to inhibit glioblastoma invasion and growth in several experimental models, and dapsone has been repeatedly been shown to clinically inhibit IL-8 function when used to treat human neutrophilic dermatoses, we believe that dapsone thereby reduces seizures by countering IL-8 function and may similarly retard glioblastoma growth by such anti-IL-8 function.
Sigma-Aldrich
4-Aminophenyl sulfone, 97%
Supelco
Dapsone, VETRANAL®, analytical standard
Dapsone, European Pharmacopoeia (EP) Reference Standard