MilliporeSigma
  • Finasteride-related Leydig cell tumour: report of a case and literature review.
Finasteride-related Leydig cell tumour: report of a case and literature review.
Andrologia (2011-09-29)
D Berthold, B Lhermitte, M Uffer, A Doerfler
ABSTRACT
Leydig cell tumours (LCTs) of the testis are rare. Their origin is still unknown. This case report describes a potential relationship between LCT and prolonged exposure to Finasteride.
Sigma-Aldrich
Finasteride, ≥98% (HPLC), powder
Pricing and availability is not currently available.
Finasteride, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Finasteride for peak identification, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.