MilliporeSigma
  • A review of the potential benefits of pentoxifylline in diabetic and non-diabetic proteinuria.
A review of the potential benefits of pentoxifylline in diabetic and non-diabetic proteinuria.
Journal of pharmacy & pharmaceutical sciences : a publication of the Canadian Society for Pharmaceutical Sciences, Societe canadienne des sciences pharmaceutiques (2011-04-20)
Shirinsadat Badri, Simin Dashti-Khavidaki, Mahboob Lessan-Pezeshki, Mohammad Abdollahi
ABSTRACT
Chronic kidney disease (CKD) as a considerable health problem may have proteinuria as the main complication and strong risk factor to reach end-stage renal disease (ESRD). Decreasing proteinuria is the mainstay of therapy in order to delay the progression of CKD. Current therapeutic regimens provide only partial renoprotection, and a substantial number of patients who have proteinuria progress to ESRD. Pentoxifylline (PTF) is known for its potent inhibitory effects against cell proliferation and inflammation which play important roles in CKD progression. Data derived from both human studies and animal models demonstrated that PTF has broad-spectrum renoprotective effects and therefore, provide a scientific basis for the use of PTF as an anti-proteinuric agent. Conclusion of this review is that short-term use of PTF may produce a significant reduction of proteinuria in subjects with diabetic and also non-diabetic kidney diseases but the reports of long-term use of PTF also show that urinary protein excretion exhibits a progressive and sustained reduction in patients treated with PTF. Whether the long-term use of PTF could be a pharmacological alternative for delaying or preventing the development of end stage renal disease, is among the questions that remained to be appropriately answered in large-scale clinical trials.
Sigma-Aldrich
Pentoxifylline, solid
Pentoxifylline, European Pharmacopoeia (EP) Reference Standard
USP
Pentoxifylline, United States Pharmacopeia (USP) Reference Standard