MilliporeSigma
Imaging evaluation of intrathecal baclofen pump-catheter systems.
AJNR. American journal of neuroradiology (2010-10-30)
A C Miracle, M A Fox, R N Ayyangar, A Vyas, S K Mukherji, D J Quint
ABSTRACT
ITB pumps are widely used in the treatment of intractable spasticity for many clinical indications, including cerebral palsy and spinal cord injury. High-dose intrathecal administration places the patient at significant risk for withdrawal in the event of device malfunction, necessitating rapid and complete evaluation of the pump-catheter system. This article reviews the approach to imaging evaluation of ITB pump-catheter systems, with specific emphasis on radiography, fluoroscopy, CT, and nuclear scintigraphy.
Sigma-Aldrich
(±)-Baclofen, ≥98% (HPLC), solid
Baclofen, European Pharmacopoeia (EP) Reference Standard