MilliporeSigma
  • [Advanced achievements about neuroprotective mechanisms of paeoniflorin].
[Advanced achievements about neuroprotective mechanisms of paeoniflorin].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2010-09-09)
Yefang Zhu, Shanshan Dang, Ziyu Hua
ABSTRACT
Paeoniflorin is one of the bioactive components of Paeonia lactiflora, a traditional Chinese herbal medicine. Some recent studies prove its distinguished neuroprotective effect. These neuroprotective mechanisms have become hot points and show closely correlated to activating adenosine A1 receptor, ameliorating the function of cholinergic nerve, regulating ion channel homeostasis, retarding oxidative stress and apoptosis of the neurocytes, promoting nerve growth, having an influence on astrocytes and being ableto penetrate though blood brain barrier. In this review, we present the neuroprotective mechanisms of paeoniflorin in the following eight aspects.
Sigma-Aldrich
Paeoniflorin, ≥98% (HPLC)
Pricing and availability is not currently available.
Supelco
Paeoniflorin, analytical standard
Pricing and availability is not currently available.